C12H316CE8C6 Chemcia Scientific, LLC

2-Phenyl-5,8-dihydro-6H-pyrido[3,4-d]pyrimidine-7-carboxylic acid tert-butyl ester

  • MDL number MFCD20278063
  • Cas Number : 1279816-05-5
  • Molecular Weight : 311.38697

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1511

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1511
Availability details for
Quantity
  • Availability
  • Add to Cart