C12H316CE8C7 Chemcia Scientific, LLC

2-Phenyl-5,7-dihydro-pyrrolo[3,4-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD22565690
  • Cas Number : 1395493-09-0
  • Molecular Weight : 297.35988

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1513

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1513
Availability details for
Quantity
  • Availability
  • Add to Cart