C12H316CE8CA Chemcia Scientific, LLC

6-Bromo-3-iodo-1H-quinolin-4-one

  • MDL number MFCD19981872
  • Cas Number : 1320361-71-4
  • Molecular Weight : 349.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-iodo-4(1H)-quinolinone
Inchi Code 1S/C9H5BrINO/c10-5-1-2-8-6(3-5)9(13)7(11)4-12-8/h1-4H,(H,12,13)
Inchi Key RTVKEUFMIJMMNH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1524

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1524
Availability details for
Quantity
  • Availability
  • Add to Cart