C12H316CE8CE Chemcia Scientific, LLC

3-Bromo-4-oxo-1,4-dihydro-quinoline-6-carboxylic acid methyl ester

  • MDL number MFCD22565699
  • Cas Number : 1395493-07-8
  • Molecular Weight : 282.09531

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1530

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1530
Availability details for
Quantity
  • Availability
  • Add to Cart