C12H316CE8D2 Chemcia Scientific, LLC

3-Bromo-6-chloro-1H-quinolin-4-one

  • MDL number MFCD22565707
  • Cas Number : 860230-86-0
  • Molecular Weight : 258.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-6-chloro-4-quinolinol
Inchi Code 1S/C9H5BrClNO/c10-7-4-12-8-2-1-5(11)3-6(8)9(7)13/h1-4H,(H,12,13)
Inchi Key MGAGUVXHIWPHRK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1540

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1540
Availability details for
Quantity
  • Availability
  • Add to Cart