C12H316CE8DD Chemcia Scientific, LLC

6-Nitro-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid

  • MDL number MFCD02169438
  • Cas Number : 52980-08-2
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxy-6-nitro-2-quinolinecarboxylic acid
Inchi Code 1S/C10H6N2O5/c13-9-4-8(10(14)15)11-7-2-1-5(12(16)17)3-6(7)9/h1-4H,(H,11,13)(H,14,15)
Inchi Key KQFRZQHCZNAUOS-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1558

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1558
Availability details for
Quantity
  • Availability
  • Add to Cart