C12H316CE8DF Chemcia Scientific, LLC

8-Bromo-6-chloro-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09836172
  • Cas Number : 1000017-98-0
  • Molecular Weight : 275.49028

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1560

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1560
Availability details for
Quantity
  • Availability
  • Add to Cart