C12H316CE8E5 Chemcia Scientific, LLC

2-Methylsulfanyl-7,8-dihydro-5H-pyrido[4,3-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD21604165
  • Cas Number : 259809-78-4
  • Molecular Weight : 281.37928

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1573

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1573
Availability details for
Quantity
  • Availability
  • Add to Cart