C12H316CE8E7 Chemcia Scientific, LLC

6-Methoxy-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid

  • MDL number MFCD09946644
  • Cas Number : 52980-06-0
  • Molecular Weight : 219.19868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1582

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1582
Availability details for
Quantity
  • Availability
  • Add to Cart