C12H316CE8ED Chemcia Scientific, LLC

4-Chloro-3-iodo-cinnoline

  • MDL number MFCD22565718
  • Cas Number : 82452-99-1
  • Molecular Weight : 290.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-3-iodocinnoline
Inchi Code 1S/C8H4ClIN2/c9-7-5-3-1-2-4-6(5)11-12-8(7)10/h1-4H
Inchi Key DEOSDIOOYNWSMJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1589

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1589
Availability details for
Quantity
  • Availability
  • Add to Cart