C12H316CE8F6 Chemcia Scientific, LLC

4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD07434961
  • Cas Number : 23443-10-9
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4H-pyrido[1,2-a]pyrimidin-4-one
Inchi Code 1S/C8H6N2O/c11-8-4-5-9-7-3-1-2-6-10(7)8/h1-6H
Inchi Key NYJWYCAHJRGKMI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1601

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1601
Availability details for
Quantity
  • Availability
  • Add to Cart