C12H316CE8F7 Chemcia Scientific, LLC

8-Methyl-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD22565720
  • Cas Number : 85653-81-2
  • Molecular Weight : 160.17691

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1602

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1602
Availability details for
Quantity
  • Availability
  • Add to Cart