C12H316CE8FA Chemcia Scientific, LLC

7-Chloro-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD07434964
  • Cas Number : 517895-69-1
  • Molecular Weight : 180.59485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1607

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1607
Availability details for
Quantity
  • Availability
  • Add to Cart