C12H316CE8FB Chemcia Scientific, LLC

2-Methyl-quinoline-4-carboxylic acid methyl ester

  • MDL number MFCD01961559
  • Cas Number : 55625-40-6
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methyl-4-quinolinecarboxylate
Inchi Code 1S/C12H11NO2/c1-8-7-10(12(14)15-2)9-5-3-4-6-11(9)13-8/h3-7H,1-2H3
Inchi Key IMGVYHBEXUHRHX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Gray Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1611

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1611
Availability details for
Quantity
  • Availability
  • Add to Cart