C12H316CE905 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride

  • MDL number MFCD26792521
  • Cas Number : 1187830-47-2
  • Molecular Weight : 159.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5,6,7-tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride
Inchi Code 1S/C6H9N3.ClH/c1-2-5-6(7-3-1)4-8-9-5;/h4,7H,1-3H2,(H,8,9);1H
Inchi Key PVKOWBFBILDXPG-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1804

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1804
Availability details for
Quantity
  • Availability
  • Add to Cart