C12H316CE908 Chemcia Scientific, LLC

1-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride

  • MDL number MFCD26792487
  • Molecular Weight : 173.64679

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1808

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1808
Availability details for
Quantity
  • Availability
  • Add to Cart