C12H316CE90B Chemcia Scientific, LLC

7,8-Dihydro-6H-pyrido[3,2-d]pyrimidine-5-carboxylic acid tert-butyl ester

  • MDL number MFCD15146665
  • Cas Number : 1174007-44-3
  • Molecular Weight : 235.28819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1814

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1814
Availability details for
Quantity
  • Availability
  • Add to Cart