C12H316CE90E Chemcia Scientific, LLC

2-Methanesulfonyl-7,8-dihydro-6H-pyrido[3,2-d]pyrimidine-5-carboxylic acid tert-butyl ester

  • MDL number MFCD23115395
  • Cas Number : 1421311-97-8
  • Molecular Weight : 313.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-(methylsulfonyl)-7,8-dihydropyrido[3,2-d]pyrimidine-5(6H)-carboxylate
Inchi Code 1S/C13H19N3O4S/c1-13(2,3)20-12(17)16-7-5-6-9-10(16)8-14-11(15-9)21(4,18)19/h8H,5-7H2,1-4H3
Inchi Key WGGHFJSULDYZFW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1826

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1826
Availability details for
Quantity
  • Availability
  • Add to Cart