C12H316CE911 Chemcia Scientific, LLC

4-Bromo-3-iodo-benzoic acid methyl ester

  • MDL number MFCD18398802
  • Cas Number : 1257231-51-8
  • Molecular Weight : 340.94422

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2029

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2029
Availability details for
Quantity
  • Availability
  • Add to Cart