C12H316CE912 Chemcia Scientific, LLC

4-Chloro-3-iodo-benzoic acid methyl ester

  • MDL number MFCD09263357
  • Cas Number : 365526-34-7
  • Molecular Weight : 296.49322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2030

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2030
Availability details for
Quantity
  • Availability
  • Add to Cart