C12H316CE913 Chemcia Scientific, LLC

4-Fluoro-3-iodo-benzoic acid methyl ester

  • MDL number MFCD11870177
  • Cas Number : 1121586-29-5
  • Molecular Weight : 280.03862

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2032

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2032
Availability details for
Quantity
  • Availability
  • Add to Cart