C12H316CE915 Chemcia Scientific, LLC

(2-Amino-5,6,7,8-tetrahydro-quinazolin-6-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD24843107
  • Cas Number : 1073633-86-9
  • Molecular Weight : 264.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (2-amino-5,6,7,8-tetrahydroquinazolin-6-yl)carbamate
Inchi Code 1S/C13H20N4O2/c1-13(2,3)19-12(18)16-9-4-5-10-8(6-9)7-15-11(14)17-10/h7,9H,4-6H2,1-3H3,(H,16,18)(H2,14,15,17)
Inchi Key ADZQBEXWLVQODS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2042

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2042
Availability details for
Quantity
  • Availability
  • Add to Cart