C12H316CE918 Chemcia Scientific, LLC

3-Dimethylaminomethylene-4-oxo-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD24843110
  • Cas Number : 727382-73-2
  • Molecular Weight : 288.3494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2048

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2048
Availability details for
Quantity
  • Availability
  • Add to Cart