C12H316CE919 Chemcia Scientific, LLC

2-Amino-7,8-dihydro-5H-pyrido[4,3-d]pyrimidine-6-carboxylic acid benzyl ester

  • MDL number MFCD24843113
  • Cas Number : 1440526-34-0
  • Molecular Weight : 284.32037

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2051

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2051
Availability details for
Quantity
  • Availability
  • Add to Cart