C12H316CE91C Chemcia Scientific, LLC

2-Amino-7,8-dihydro-6H-quinazolin-5-one

  • MDL number MFCD00731537
  • Cas Number : 21599-36-0
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-7,8-dihydro-5(6H)-quinazolinone
Inchi Code 1S/C8H9N3O/c9-8-10-4-5-6(11-8)2-1-3-7(5)12/h4H,1-3H2,(H2,9,10,11)
Inchi Key ZXWJMLMXKMSYTP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2058

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2058
Availability details for
Quantity
  • Availability
  • Add to Cart