C12H316CE91E Chemcia Scientific, LLC

(2-Amino-benzothiazol-6-yl)-acetic acid ethyl ester

  • MDL number MFCD05664559
  • Cas Number : 68195-02-8
  • Molecular Weight : 236.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (2-amino-1,3-benzothiazol-6-yl)acetate
Inchi Code 1S/C11H12N2O2S/c1-2-15-10(14)6-7-3-4-8-9(5-7)16-11(12)13-8/h3-5H,2,6H2,1H3,(H2,12,13)
Inchi Key HBYRGKVSKZCQGF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2067

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2067
Availability details for
Quantity
  • Availability
  • Add to Cart