C12H316CE920 Chemcia Scientific, LLC

(2-Bromo-benzothiazol-6-yl)-acetic acid

  • MDL number MFCD19687422
  • Cas Number : 933742-71-3
  • Molecular Weight : 272.12167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2074

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2074
Availability details for
Quantity
  • Availability
  • Add to Cart