C12H316CE924 Chemcia Scientific, LLC

(6-Bromo-benzothiazol-2-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD16036619
  • Cas Number : 1244041-71-1
  • Molecular Weight : 329.21761

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2082

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2082
Availability details for
Quantity
  • Availability
  • Add to Cart