C12H316CE933 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-2H-pyrazolo[4,3-c]pyridin-3-ol dihydrochloride

  • MDL number MFCD21647877
  • Cas Number : 1260898-28-9
  • Molecular Weight : 212.08007

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data 2HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2202

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2202
Availability details for
Quantity
  • Availability
  • Add to Cart