C12H316CE935 Chemcia Scientific, LLC

1,2,4,5,6,7-Hexahydro-indazol-3-one

  • MDL number MFCD01407953
  • Cas Number : 4344-73-4
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5,6,7-tetrahydro-1H-indazol-3-ol
Inchi Code 1S/C7H10N2O/c10-7-5-3-1-2-4-6(5)8-9-7/h1-4H2,(H2,8,9,10)
Inchi Key UFNPJQRQWKZJNL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2207

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2207
Availability details for
Quantity
  • Availability
  • Add to Cart