C12H316CE936 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-2H-indazol-3-ol

  • MDL number MFCD00462221
  • Cas Number : 402602-24-8
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5,6,7-tetrahydro-2H-indazol-3-ol
Inchi Code 1S/C7H10N2O/c10-7-5-3-1-2-4-6(5)8-9-7/h1-4H2,(H2,8,9,10)
Inchi Key UFNPJQRQWKZJNL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2208

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2208
Availability details for
Quantity
  • Availability
  • Add to Cart