C12H316CE93B Chemcia Scientific, LLC

6-Fluoro-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD09994901
  • Cas Number : 881841-32-3
  • Molecular Weight : 164.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoroimidazo[1,2-a]pyridine-2-carbaldehyde
Inchi Code 1S/C8H5FN2O/c9-6-1-2-8-10-7(5-12)4-11(8)3-6/h1-5H
Inchi Key PHQLIVRLJJGESN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2214

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2214
Availability details for
Quantity
  • Availability
  • Add to Cart