C12H316CE941 Chemcia Scientific, LLC

7-Methyl-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD10699196
  • Cas Number : 4999-42-2
  • Molecular Weight : 133.15394

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2231

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2231
Availability details for
Quantity
  • Availability
  • Add to Cart