C12H316CE947 Chemcia Scientific, LLC

2-Chloro-benzothiazole-6-carboxylic acid methyl ester

  • MDL number MFCD13190216
  • Cas Number : 90792-69-1
  • Molecular Weight : 227.67067

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2238

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2238
Availability details for
Quantity
  • Availability
  • Add to Cart