C12H316CE957 Chemcia Scientific, LLC

2-Bromo-4-methoxy-benzothiazole

  • MDL number MFCD08459038
  • Cas Number : 3622-39-7
  • Molecular Weight : 244.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-methoxy-1,3-benzothiazole
Inchi Code 1S/C8H6BrNOS/c1-11-5-3-2-4-6-7(5)10-8(9)12-6/h2-4H,1H3
Inchi Key ZVCFJHNSAMCWGM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2285

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2285
Availability details for
Quantity
  • Availability
  • Add to Cart