C12H316CE95C Chemcia Scientific, LLC

2-Amino-4-methoxy-nicotinic acid

  • MDL number MFCD06208729
  • Cas Number : 773099-92-6
  • Molecular Weight : 168.15341

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2344

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2344
Availability details for
Quantity
  • Availability
  • Add to Cart