C12H316CE95E Chemcia Scientific, LLC

4-Chloro-quinoline-8-carbonitrile

  • MDL number MFCD08063206
  • Cas Number : 132664-45-0
  • Molecular Weight : 188.61775

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2346

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2346
Availability details for
Quantity
  • Availability
  • Add to Cart