C12H316CE960 Chemcia Scientific, LLC

4-Chloro-2-fluoro-5-iodo-benzoic acid methyl ester

  • MDL number MFCD24107097
  • Cas Number : 1516210-03-9
  • Molecular Weight : 314.48365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2350

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2350
Availability details for
Quantity
  • Availability
  • Add to Cart