C12H316CE964 Chemcia Scientific, LLC

8-Bromo-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD11504929
  • Cas Number : 942227-30-7
  • Molecular Weight : 282.09531

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2358

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2358
Availability details for
Quantity
  • Availability
  • Add to Cart