C12H316CE968 Chemcia Scientific, LLC

2,8-Dibromo-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD22628138
  • Cas Number : 1257705-07-9
  • Molecular Weight : 276.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,8-dibromo-[1,2,4]triazolo[1,5-a]pyridine
Inchi Code 1S/C6H3Br2N3/c7-4-2-1-3-11-5(4)9-6(8)10-11/h1-3H
Inchi Key LSTOPRBLSYPTFQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2372

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2372
Availability details for
Quantity
  • Availability
  • Add to Cart