C12H316CE96A Chemcia Scientific, LLC

2,5-Dibromo-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD20527883
  • Cas Number : 1214901-64-0
  • Molecular Weight : 276.91891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2374

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2374
Availability details for
Quantity
  • Availability
  • Add to Cart