C12H316CE96E Chemcia Scientific, LLC

2-Bromo-8-chloro-6-methyl-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD26131657
  • Molecular Weight : 246.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-8-chloro-6-methyl-[1,2,4]triazolo[1,5-a]pyridine
Inchi Code 1S/C7H5BrClN3/c1-4-2-5(9)6-10-7(8)11-12(6)3-4/h2-3H,1H3
Inchi Key QADFAKOHTXIJDT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2378

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2378
Availability details for
Quantity
  • Availability
  • Add to Cart