C12H316CE96F Chemcia Scientific, LLC

8-Bromo-imidazo[1,2-a]pyridin-6-ylamine hydrochloride

  • MDL number MFCD22192409
  • Cas Number : 1373233-09-0
  • Molecular Weight : 248.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromoimidazo[1,2-a]pyridin-6-amine hydrochloride
Inchi Code 1S/C7H6BrN3.ClH/c8-6-3-5(9)4-11-2-1-10-7(6)11;/h1-4H,9H2;1H
Inchi Key DUAYTYVYTSNOFJ-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2380

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2380
Availability details for
Quantity
  • Availability
  • Add to Cart