C12H316CE973 Chemcia Scientific, LLC

2-(4-Bromo-phenyl)-2-methyl-propionic acid

  • MDL number MFCD02684220
  • Cas Number : 32454-35-6
  • Molecular Weight : 243.10197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2385

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2385
Availability details for
Quantity
  • Availability
  • Add to Cart