C12H316CE976 Chemcia Scientific, LLC

[2-(3-Formyl-benzoylamino)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD26131663
  • Cas Number : 1820609-00-4
  • Molecular Weight : 292.33765

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2392

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2392
Availability details for
Quantity
  • Availability
  • Add to Cart