C12H316CE97F Chemcia Scientific, LLC

(6-Methoxy-pyridin-3-yl)-methyl-amine

  • MDL number MFCD10697832
  • Cas Number : 61771-67-3
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2403

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2403
Availability details for
Quantity
  • Availability
  • Add to Cart