C12H316CE980 Chemcia Scientific, LLC

(6-Methoxy-pyridin-3-yl)-dimethyl-amine

  • MDL number MFCD18802420
  • Cas Number : 342793-50-4
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(6-methoxy-3-pyridinyl)-N,N-dimethylamine
Inchi Code 1S/C8H12N2O/c1-10(2)7-4-5-8(11-3)9-6-7/h4-6H,1-3H3
Inchi Key CREDJITUIHHIFL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2404

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2404
Availability details for
Quantity
  • Availability
  • Add to Cart