C12H316CE983 Chemcia Scientific, LLC

3-Iodo-imidazo[1,2-a]pyridine-8-carboxylic acid methyl ester

  • MDL number MFCD06739271
  • Cas Number : 885276-95-9
  • Molecular Weight : 302.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-iodoimidazo[1,2-a]pyridine-8-carboxylate
Inchi Code 1S/C9H7IN2O2/c1-14-9(13)6-3-2-4-12-7(10)5-11-8(6)12/h2-5H,1H3
Inchi Key OXTNGJDEOUMONG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2435

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2435
Availability details for
Quantity
  • Availability
  • Add to Cart