C12H316CE984 Chemcia Scientific, LLC

3-Iodo-6-nitro-imidazo[1,2-a]pyridine

  • MDL number MFCD07374358
  • Cas Number : 886371-73-9
  • Molecular Weight : 289.03323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2436

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2436
Availability details for
Quantity
  • Availability
  • Add to Cart