C12H316CE986 Chemcia Scientific, LLC

3-Iodo-8-methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD24199924
  • Cas Number : 1545535-05-4
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-iodo-8-methylimidazo[1,2-a]pyridine
Inchi Code 1S/C8H7IN2/c1-6-3-2-4-11-7(9)5-10-8(6)11/h2-5H,1H3
Inchi Key LSKAHQYWBXCYBD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2438

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2438
Availability details for
Quantity
  • Availability
  • Add to Cart